BR708728
BRAND® disposable BLAUBRAND® micropipettes, intraMark
tube volume 20-40 μL , Dual marks, red
Synonym(s):
Micropipettes, Pipettes, intraMARK
Select a Size
1000 EA
€139.00
€139.00
Please contact Customer Service for Availability
All Photos(1)
Select a Size
Change View
1000 EA
€139.00
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
€139.00
Please contact Customer Service for Availability
material
red
feature
DE-M marking, To Contain
packaging
pack of 1000 ea (4 boxes of 250)
manufacturer/tradename
BRAND 708728
L
125 mm
tube volume
20-40 μL , Dual marks
General description
DIN ISO 7550. DE-M marking for volumes greater than 5 µl. Calibrated to contain (TC, In). Color code to ISO for clear identification.
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)