Key Documents
BR701372
BRAND® sealing film for microplates
self-adhesive, suitable for (for luminescence measurement), vinyl
Synonym(s):
Accessories, PCR, Plates, Sealing films
Select a Size
50 EA
€53.30
€53.30
Please contact Customer Service for Availability
Select a Size
Change View
50 EA
€53.30
About This Item
UNSPSC Code:
41122410
NACRES:
NB.22
€53.30
Please contact Customer Service for Availability
material
vinyl
vinyl film
white
sterility
non-sterile
feature
self-adhesive
packaging
pack of 50 ea
manufacturer/tradename
BRAND 701372
suitability
suitable for (for luminescence measurement)
General description
White and black films provide support for fluorescence and luminescence measurements and for microscopy. Weak signals are amplified and interference of light from outside is minimized.
Features and Benefits
  • Black, light-absorbing film for optimal fluorescence measurements
  • White, light-reflecting film for amplified luminescent signal
  • Storable down to -40 °C
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)