Skip to Content
Merck
All Photos(1)

Key Documents

BR701360

BRAND® mat cover for deep well plate

polyethylene, volume 1.1 mL, size 96 wells

Synonym(s):

Accessories, Cover mats, Deep-well Plates, Sample Storage

Sign Into View Organizational & Contract Pricing

Select a Size

24 EA
€79.80

€79.80


Please contact Customer Service for Availability


Select a Size

Change View
24 EA
€79.80

About This Item

UNSPSC Code:
41122102
NACRES:
NB.22

€79.80


Please contact Customer Service for Availability

material

polyethylene

sterility

non-sterile

manufacturer/tradename

BRAND 701360

size

1.2 mL , elevated skirt
96 wells

volume

1.1 mL

Looking for similar products? Visit Product Comparison Guide

General description

These cover mats reduce the maximum volume of wells but minimize evaporation.They can be used in conjunction with adhesive sealing films for added protection and containment.

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service