Skip to Content
Merck
All Photos(1)

Key Documents

BR701346

BRAND® 96-well deep well plate, stackable

volume 0.5 mL, polypropylene

Synonym(s):

Deep-well Plates, Sample Storage

Sign Into View Organizational & Contract Pricing

Select a Size

48 EA
€226.00

€226.00


Please contact Customer Service for Availability


Select a Size

Change View
48 EA
€226.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
material:
polypropylene
round bottom wells
size:
96 well
sterility:
non-sterile
binding type:
non-treated surface
feature:
skirt

€226.00


Please contact Customer Service for Availability

material

polypropylene
round bottom wells

description

alphanumeric identification system

sterility

non-sterile

feature

skirt

packaging

pack of 48 ea

manufacturer/tradename

BRAND 701346

size

96 well

volume

0.5 mL

working volume

500 μm

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • BRAND® 96-well deep well plates are made of high-grade chemically resistant polypropylene.
  • Ideal for use in various automated processes including High-Throughput Screening (HTS), cell and tissue cultures, serial dilutions, reagent transfer, and sample storage down to -80 °C.
  • The raised edges of the well facilitate a secure seal and prevent sample contamination.
  • Stackable - A convenient space-saving option.
  • ′U′ bottom base ensures optimal sample mixing and collection.
  • Alphanumeric coding for easy identification.
  • Free of DNA, DNAse and RNAse for sample integrity.

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service