Key Documents
BR6638
BRAND® filling valve for Dispensette® III and Dispensette® Organic
for 25-100 mL
Synonym(s):
Accessories, Bottle-top Dispenser, Dispenser, Dispensette®, Filling valve
About This Item
UNSPSC Code:
40141616
packaging
pack of 1 ea
manufacturer/tradename
BRAND 6638
General description
With olive-shaped nozzle (PEEK) and sealing ring (FKM).
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)