Key Documents
BR61605
BRAND® silicone paste UNISILKON TK M 1011 tube of 50 g
Select a Size
1 EA
€35.20
€35.20
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€35.20
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
€35.20
Please contact Customer Service for Availability
packaging
pack of 1 ea
manufacturer/tradename
BRAND 61605
Related Categories
General description
The BRAND® silicone paste UNISILKON TK M 1011 comes in a convenient 50 g tube and features a low viscosity formulation, making it ideal for various applications. With an operating temperature range of -40 to 160 °C, this silicone paste is highly versatile and suitable for use in extreme conditions. It is specifically designed for greasing ground joints, ensuring a smooth and effective seal.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)