BR462512
BRAND® rubber conical gasket for filter funnels and filter flasks
size 22 mm
Synonym(s):
Filter, Rubber gasket
All Photos(1)
About This Item
Recommended Products
material
EPDM rubber
packaging
pack of 10 ea
manufacturer/tradename
BRAND 462512
size
22 mm
Related Categories
General description
Conical gaskets to place between filter funnel and filter flask.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG