Key Documents
BR43228
BRAND® density bottle, Gay-Lussac pattern, uncalibrated
capacity 50 mL, borosilicate glass
Synonym(s):
Gay-Lussac
Select a Size
2 EA
€82.90
€82.90
Please contact Customer Service for Availability
Select a Size
Change View
2 EA
€82.90
About This Item
UNSPSC Code:
41121800
NACRES:
NB.71
€82.90
Please contact Customer Service for Availability
material
borosilicate glass
glass stopper
narrow-mouth bottle
round bottle (Gay-Lussac)
white bottle
Joint
joint ST/NS neck10/19
feature
with cap
packaging
pack of 2 ea
manufacturer/tradename
BRAND 43228
capacity
50 mL
General description
DIN ISO 3507, type Gay-Lussac. Stopper NS 10/19 with capillary. Top of stopper ground and polished. Nominal capacity printed on the bottom.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Protocols
Density bottles determine liquid density, calibrated 'to contain' for accuracy, though not volumetric instruments like volumetric flasks.