BR43038
BRAND® glass bottle-top tilt pipette dispenser
volume 100 mL, with bottle
Synonym(s):
Automatic pipettes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
Recommended Products
feature
Calibrated to deliver (TD, Ex)
with bottle
manufacturer/tradename
BRAND 43038
volume
100 mL
color
green markings
fitting
(29/32)
Looking for similar products? Visit Product Comparison Guide
General description
Calibrated to deliver (TD, Ex). Inscriptions in green enamel. The automatic dispenser is particularly well-suited for dispensing suspensions. Delivery range approximately +/- 10%.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service