BR152800
BRAND® tubing connector, T-shape
polypropylene, for tubing i.d., 3 ‑ 4 mm
Synonym(s):
Tubing, Tubing Connectors
Select a Size
€21.70
Available to ship on24 April 2025Details
Select a Size
About This Item
€21.70
Available to ship on24 April 2025Details
Recommended Products
material
polypropylene
feature
for tubing i.d. 3 ‑ 4 mm
packaging
pack of 20 ea
manufacturer/tradename
BRAND 152800
L
20 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service