Key Documents
BR150040
BRAND® watch glass
soda-lime glass, diam. 80 mm , ground edges stress-reduced
Synonym(s):
Dishes, Watch glasses
Select a Size
10 EA
€32.00
€32.00
Please contact Customer Service for Availability
Select a Size
Change View
10 EA
€32.00
About This Item
UNSPSC Code:
41121801
NACRES:
NB.43
€32.00
Please contact Customer Service for Availability
material
soda-lime glass
manufacturer/tradename
BRAND 150040
diam.
80 mm , ground edges stress-reduced
General description
The BRAND® watch glass is made from soda-lime glass and complies with DIN 12341 standards. It features ground edges for safe handling and a refined finish. This watch glass is stress-reduced to minimize the risk of shattering, enhancing safety during laboratory use. Ideal for covering beakers or evaporating dishes, it provides an effective solution for preventing contamination and evaporation of samples. Its durable design makes it a reliable tool in various laboratory applications, ensuring optimal performance in your experiments.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)