Skip to Content
Merck
All Photos(1)

Key Documents

BMSE4100E

Benchmark SmartBlue Mini Transilluminator

AC/DC input 230 V, EuroPlug

Synonym(s):

blue light, gel imaging, gel casting, transillumination

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€529.00

€529.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EACH
€529.00

About This Item

UNSPSC Code:
41106000
NACRES:
NB.13

€529.00


Please contact Customer Service for Availability

manufacturer/tradename

Benchmark Scientific E4100-E

Looking for similar products? Visit Product Comparison Guide

General description

Safer for samples, safer for you: Accuris SmartBlueMini Blue Light Transilluminator is the perfect partner for most common safe DNA stains. It emits blue light at a wavelength of 465nm, perfect for excitation of most green fluorophors, including our Accuris’ SmartGlow, SYBR Green, SYBR Safe, SYBR Gold, GelStar, GelGreen, to name a few. The SmartBlue Mini also works for excitation of EtBr (though fluorescence is not as bright as with UV), see our Technical Bulletin with sample images. Exposing DNA to blue wavelength does not cause molecular damage, and results in a much higher downstream cloning efficiency as compared to even short UV light exposures. In addition, the visible blue light is not damaging to the user’s skin and is less harmful to the eyes.*

Application

Gel documentation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service