Merck
All Photos(4)
Documents
BAH327870000
SP Bel-Art® Carboy Pump
for use with 20 L (5 Gal) carboy
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H32787-0000
General description
The Poly-Hand Pump threads directly to a standard drum bung or carboy. It is constructed entirely of rugged polyethylene and is inert to strong acids, alkalis and many solvents.
Features and Benefits
  • Delivers by positive pressure on the downstroke of the piston; does not pressurize the vessel
  • Ergonomic ‘T’ handle fits comfortably in a gloved hand
  • Pump will efficiently deliver against a 3.65m (12ft) head of water and empty a drum or carboy to within 3 mm (⅛”) of the bottom
  • Sizes are available to fit 114L and 220L (30/50gal) plastic or steel drums and 20L (5gal) carboy (F10795-0000)
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP