Skip to Content
Merck
All Photos(3)

Key Documents

BAH131941011

SP Bel-Art® Benchtop Disposal Can

with motion sensor lid

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€68.80

€68.80


Please contact Customer Service for Availability


Select a Size

Change View
1 EACH
€68.80

About This Item

UNSPSC Code:
41122800
NACRES:
NB.43

€68.80


Please contact Customer Service for Availability

manufacturer/tradename

Bel-Art H13194-1011

capacity

1.5 L

diam. × H

4 in. (124 mm) × 6 in. (170 mm)

General description

Compact benchtop disposal can contain odors and is perfect for small items such as pipette tips and microcentrifuge tubes.

Features and Benefits

  • Automatic opening lid provides hands free use; closes automatically and securely for a more sanitary environment
  • Holds a 25 x 25cm (10 x 10") autoclavable biohazard disposal bag (F13166-0000 or H13182-1010)
  • Includes a polypropylene liner with handle for easy emptying of contents
  • Aesthetic and durable; stainless steel body and chrome lid are resistant to bleach and easy to clean
  • Requires (3) AAA batteries (not included)
  • CE & RoHS

Legal Information

Bel-Art is a registered trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service