Key Documents
BAF378750620
SP Bel-Art® Sterileware Single Slot Powder Sampler
capacity × L 50 mL (1¾ oz) × 24.375 in. (62 cm), pack of 20 ea (individually wrapped)
Synonym(s):
powder sampler
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
material
high-density polyethylene
packaging
pack of 20 ea (individually wrapped)
manufacturer/tradename
Bel-Art F37875-0620
capacity × L
50 mL (1¾ oz) × 24.375 in. (62 cm)
General description
These sterile powder samplers are ideal for taking a single, targeted 50ml sample from free-flowing powders and granules.
Features and Benefits
  • After insertion, rotate tube to open then close the collection slot, and remove to retrieve the sample
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • High density polyethylene (HDPE) sampler heat sealed in individual polyethylene bags
  • Gamma irradiated sterile (SAL 10^-6)
  • Lot stamped for accurate tracing
Note: Sterilized products are not returnable.
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Sterileware is a trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)