Merck
All Photos(8)
Key Documents
BAF371110158
SP Bel-Art® Spinbar® magnetic stir bar
cylindrical white PTFE-coated, L × diam. 1.625 in. (41.3 mm) × 0.375 in. (9.5 mm)
Synonym(s):
Spinbar® magnetic stir bar
About This Item
UNSPSC Code:
41122800
NACRES:
NB.51
material
cylindrical white PTFE-coated
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F37111-0158
L × diam.
1.625 in. (41.3 mm) × 0.375 in. (9.5 mm)
General description
  • Spinbar® magnetic stir bar is used in conjunction with a magnetic stirring plate for uniform, consistent, and thorough mixing of chemical solutions.
  • Alnico magnet encapsulated in durable and inert FDA grade polytetrafluoroethylene (PTFE) low friction coating.
  • Multi-faceted-shaped bars provide a greater surface area and added turbulence for more vigorous, thorough mixing.
  • Autoclave safe.
Cylindrical magnetic stirring bars offer excellent centering and smooth running characteristics.
Leakproof, seamless, polished, and contains a permanent Alnico V magnet. Egg-shape bar is designed especially for use in round-bottom vessels.
Features and Benefits
  • Cylindrical magnetic stirring bars offer excellent centering and smooth running characteristics.
  • Small removable pivot ring in the center adds to their versatility
  • Pivot ring minimizes the contact area of the bar to the vessel, reduces friction and lessens marring of plastic containers
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE coating – Superior Chemical Compatibility
  • Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
  • Autoclave safe
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Spinbar is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)