Skip to Content
Merck
All Photos(2)

Key Documents

B75506

Sigma-Aldrich

β-Bromophenetole

98%

Synonym(s):

2-Bromoethyl phenyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€235.00

€235.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€235.00

About This Item

Linear Formula:
C6H5OCH2CH2Br
CAS Number:
Molecular Weight:
201.06
Beilstein:
508290
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€235.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

98%

form

crystals

bp

144 °C/40 mmHg (lit.)

mp

31-34 °C (lit.)

SMILES string

BrCCOc1ccccc1

InChI

1S/C8H9BrO/c9-6-7-10-8-4-2-1-3-5-8/h1-5H,6-7H2

InChI key

JJFOBACUIRKUPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

149.0 °F

Flash Point(C)

65 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Son T Nguyen et al.
Bioorganic & medicinal chemistry, 23(9), 2024-2034 (2015-03-31)
Recently we described a novel pyranopyridine inhibitor (MBX2319) of RND-type efflux pumps of the Enterobacteriaceae. MBX2319 (3,3-dimethyl-5-cyano-8-morpholino-6-(phenethylthio)-3,4-dihydro-1H-pyrano[3,4-c]pyridine) is structurally distinct from other known Gram-negative efflux pump inhibitors (EPIs), such as 1-(1-naphthylmethyl)-piperazine (NMP), phenylalanylarginine-β-naphthylamide (PAβN), D13-9001, and the pyridopyrimidine derivatives. Here

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service