Skip to Content
Merck
All Photos(1)

Key Documents

B2761

Sigma-Aldrich

5-Bromoindole-2-carboxylic acid

98%

Synonym(s):

NSC 73384

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€115.00

€115.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€115.00

About This Item

Linear Formula:
C9H6NO2Br
CAS Number:
Molecular Weight:
240.05
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€115.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

98%

form

powder

storage temp.

−20°C

SMILES string

OC(=O)c1cc2cc(Br)ccc2[nH]1

InChI

1S/C9H6BrNO2/c10-6-1-2-7-5(3-6)4-8(11-7)9(12)13/h1-4,11H,(H,12,13)

InChI key

YAULOOYNCJDPPU-UHFFFAOYSA-N

Application

Reactant involved in studies of biologically active molecules including:
  • Discovery of indole inhibitors of MMP-13 for treatment of arthritic diseases
  • Synthesis of indolyl ethanones as indoleamine 2,3-dioxygenase inhibitors
  • cis-Diaminocyclohexane derivatives prepared for use as factor Xa inhibitors
  • Synthesis of tubulin polymerization inhibitors and cancer cell growth inhibitors
  • Preparation of dual PPARγ/δ agonists
  • Synthesis of chemical probes to examine the role of hFPRL1 receptor in inflammation

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shahrbanoo Jahangir et al.
Stem cell research & therapy, 11(1), 436-436 (2020-10-11)
Mesenchymal stem cells are a promising cell source for chondrogenic differentiation and have been widely used in several preclinical and clinical studies. However, they are prone to an unwanted differentiation process towards hypertrophy that limits their therapeutic efficacy. Matrix metallopeptidase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service