Skip to Content
Merck
All Photos(1)

Key Documents

B0751

Sigma-Aldrich

5-Benzyloxyindole-3-carboxaldehyde

powder

Synonym(s):

5-Benzyloxy-3-formylindole, NSC 71049

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13NO2
CAS Number:
Molecular Weight:
251.28
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

form

powder

color

yellow to brown

storage temp.

2-8°C

SMILES string

O=Cc1c[nH]c2ccc(OCc3ccccc3)cc12

InChI

1S/C16H13NO2/c18-10-13-9-17-16-7-6-14(8-15(13)16)19-11-12-4-2-1-3-5-12/h1-10,17H,11H2

InChI key

DJGNUBADRQIDNQ-UHFFFAOYSA-N

Application

Reactant for preparation of:
  • Rhodanine merocyanine dyes
  • Potential topoisomerase II inhibitors
  • Azithromycin derivatives
  • Inhibitors of PI3 kinase-α and the mammalian target of rapamycin
  • Cytotoxic agents
  • Vascular endothelial growth factor (VEGF) inhibitor
  • Derivatives of vancomycin and eremomycin
  • Benzimidazoles as potential antibacterial agents
  • Light-dependent tumor necrosis factor-α antagonists
  • Serotonin 5-HT4 Receptor agonists

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service