Skip to Content
Merck
All Photos(1)

Documents

B0626

Sigma-Aldrich

5-Benzyloxyindole-3-acetic acid

crystalline

Synonym(s):

NSC 68361

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H15NO3
CAS Number:
Molecular Weight:
281.31
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

crystalline

color

off-white to tan

storage temp.

2-8°C

SMILES string

OC(=O)Cc1c[nH]c2ccc(OCc3ccccc3)cc12

InChI

1S/C17H15NO3/c19-17(20)8-13-10-18-16-7-6-14(9-15(13)16)21-11-12-4-2-1-3-5-12/h1-7,9-10,18H,8,11H2,(H,19,20)

InChI key

GKIOPUYLJUOZHJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for preparation of:
  • Inhibitors of Gli1-mediated transcription as potential anticancer agents
  • Synthetic analogs of spider toxins

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Sugawara et al.
Biochimica et biophysica acta, 1192(2), 241-246 (1994-06-22)
The effect of membrane surface potential on the uptake of several anionic compounds by liposomes (large unilamellar vesicles), which contain various amounts of dipalmitoylphosphatidylserine (DPPS), was investigated. The uptake amount of four tested anionic compounds (cefixime, benzyloxyindoleacetic acid (BOIAA), ceftibuten

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service