Skip to Content
Merck
All Photos(1)

Key Documents

ALD00022

Sigma-Aldrich

Diethyl 2-((1-(2-oxo-2-phenylethyl)-1H-pyrrol-2-yl)methylene)malonate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21NO5
CAS Number:
Molecular Weight:
355.38
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

118 °C

storage temp.

2-8°C

SMILES string

O=C(CN1C(C=C(C(OCC)=O)C(OCC)=O)=CC=C1)C2=CC=CC=C2

InChI

1S/C20H21NO5/c1-3-25-19(23)17(20(24)26-4-2)13-16-11-8-12-21(16)14-18(22)15-9-6-5-7-10-15/h5-13H,3-4,14H2,1-2H3

InChI key

JMNYUFCXEYSRIE-UHFFFAOYSA-N

Application

Building block is employed in the enantioselective synthesis of 5, 6-dihydroindolizines through a cascade sequence initiated by (S)-(–)-α,α-Diphenyl-2-pyrrolidinemethanol trimethylsilyl ether (Aldrich 677183).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service