Skip to Content
Merck
All Photos(1)

Key Documents

A79000

Sigma-Aldrich

4-Aminoquinaldine

98%

Synonym(s):

2-Methyl-4-quinolinamine, 4-Amino-2-methylquinoline, 4-Quinaldinamine, NSC 60281

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€78.90
25 G
€284.00

€78.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€78.90
25 G
€284.00

About This Item

Empirical Formula (Hill Notation):
C10H10N2
CAS Number:
Molecular Weight:
158.20
Beilstein:
118323
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€78.90


Please contact Customer Service for Availability

Request a Bulk Order

Assay

98%

form

powder

bp

333 °C (lit.)

mp

162-166 °C (lit.)

SMILES string

Cc1cc(N)c2ccccc2n1

InChI

1S/C10H10N2/c1-7-6-9(11)8-4-2-3-5-10(8)12-7/h2-6H,1H3,(H2,11,12)

InChI key

COCFIBRMFPWUDW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ishani Deb et al.
Bioorganic & medicinal chemistry, 17(16), 5782-5790 (2009-07-31)
Based on an established 3D pharmacophore, a series of quinoline derivatives were synthesized. The opioidergic properties of these compounds were determined by a competitive binding assay using (125)I-Dynorphine, (3)H-DAMGO and (125)I-DADLE for kappa, mu, and delta receptors, respectively. Results showed
Stefania Ferrari et al.
Journal of medicinal chemistry, 54(1), 211-221 (2010-12-04)
Folate analogue inhibitors of Leishmania major pteridine reductase (PTR1) are potential antiparasitic drug candidates for combined therapy with dihydrofolate reductase (DHFR) inhibitors. To identify new molecules with specificity for PTR1, we carried out a virtual screening of the Available Chemicals

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service