Skip to Content
Merck
All Photos(1)

Key Documents

A6424

Sigma-Aldrich

Astemizole

≥99%

Synonym(s):

1-(4-Fluorobenzyl)-2-(1-[4-methoxyphenethyl]piperidin-4-yl)aminobenzimidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H31FN4O
CAS Number:
Molecular Weight:
458.57
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥99%

SMILES string

COc1ccc(CCN2CCC(CC2)Nc3nc4ccccc4n3Cc5ccc(F)cc5)cc1

Biochem/physiol Actions

Non-sedating anti-histaminergic drug. Substrate for CYP3A4,5,7. Inhibitor of Pgp-mediated drug efflux and Pgp labeling by [3H]azidopine. Potential for combination therapy with anticancer drugs such as doxorubicin in resistant leukemia.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anida Hasanovic et al.
Biomedicines, 8(8) (2020-08-06)
Adrenocortical carcinoma (ACC) presents a high risk of relapse and metastases with outcomes not improving despite extensive research and new targeted therapies. We recently showed that the Hedgehog receptor Patched is expressed in ACC, where it strongly contributes to doxorubicin
Fumimasa Nomura et al.
Journal of nanobiotechnology, 9, 39-39 (2011-09-21)
Conventional in vitro approach using human ether-a-go-go related gene (hERG) assay has been considered worldwide as the first screening assay for cardiac repolarization safety. However, it does not always oredict the potential QT prolongation risk or pro-arrhythmic risk correctly. For
K A Rao et al.
Mayo Clinic proceedings, 69(6), 589-593 (1994-06-01)
An overdose of astemizole predisposes the myocardium to ventricular dysrhythmias, including torsades de pointes. Herein we describe a case of astemizole-induced torsades de pointes ventricular tachycardia and also review previous case reports in the literature. All the patients were young
Kiem Vu et al.
Medical mycology, 48(2), 255-262 (2009-07-03)
Cryptococcus neoformans is the leading cause of fungal meningitis, a life-threatening infection that occurs predominately in immuocompromised patients. Current drug therapies are limited to amphotericin B, flucytosine and the azoles since the echinocandins have no demonstrated activity against yeast like
Tjøstil Vlaar et al.
Angewandte Chemie (International ed. in English), 51(52), 13058-13061 (2012-11-20)
O(2) in, H(2)O out: Various diamines and related bisnucleophiles readily undergo oxidative isocyanide insertion with Pd(OAc)(2) (1 mol %) as the catalyst and O(2) as the terminal oxidant to give a diverse array of medicinally relevant N heterocycles. The utility

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service