Skip to Content
Merck
All Photos(1)

Key Documents

A6057

Sigma-Aldrich

4-Azidophenacyl bromide

powder

Synonym(s):

4′-Azido-2-bromoacetophenone, 4-Azido-α-bromoacetophenone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6BrN3O
CAS Number:
Molecular Weight:
240.06
Beilstein:
1961705
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

powder

reaction suitability

reaction type: click chemistry
reagent type: cross-linking reagent

color

yellow

solubility

methanol: 50 mg/mL

storage temp.

2-8°C

SMILES string

BrCC(=O)c1ccc(cc1)N=[N+]=[N-]

InChI

1S/C8H6BrN3O/c9-5-8(13)6-1-3-7(4-2-6)11-12-10/h1-4H,5H2

InChI key

LZJPDRANSVSGOR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Photoactive, heterobifunctional cross-linking reagent. Typically, the initial reaction couples to sulfhydryl in the pH range 7.0-8.0. Second bonding occurs during UV irradiation (250 nm) via reactive nitrene. The latter bonding is rapid and non-specific.

Caution

Reducing agents such as thiols may reduce the azide to amine and should be avoided. Initial manipulations and coupling should be performed under reduced light.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Sol. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

D Hu et al.
The Journal of biological chemistry, 275(4), 2705-2712 (2000-01-25)
The PI-SceI protein is an intein-encoded homing endonuclease that initiates the mobility of its gene by making a double strand break at a single site in the yeast genome. The PI-SceI protein splicing and endonucleolytic active sites are separately located
S H Hixson et al.
Biochemistry, 14(19), 4251-4254 (1975-09-23)
The synthesis of the photochemically labile bifunctional reagent p-azidophenacyl bromide (1) is described. This compound may be covalently attached to a known site of an enzyme or other macromolecule by nucleophilic displacement at the alpha-bromo ketone moiety. Subsequent irradiation of
Nan Zhang et al.
Nucleic acids research, 37(18), 5981-5992 (2009-08-21)
sigma(54)-dependent transcription requires activation by bacterial enhancer binding proteins (bEBPs). bEBPs are members of the AAA+ (ATPases associated with various cellular activities) protein family and typically form hexameric structures that are crucial for their ATPase activity. The precise mechanism by
A Robinson et al.
The Biochemical journal, 242(3), 767-777 (1987-03-15)
An azidophenacyl derivative of a chemically synthesized consensus signal peptide has been prepared. The peptide, when photoactivated in the presence of rough or high-salt-stripped microsomes from pancreas, leads to inhibition of their activity in cotranslational processing of secretory pre-proteins translated
L M Hsu et al.
Journal of molecular biology, 172(1), 57-76 (1984-01-05)
tRNAPhe and tRNAVal of Escherichia coli were derivatized at the S4U8 position with p-azidophenacyl and p-azidophenacylacetate photoaffinity probes. The modified tRNAs could still function efficiently in all of the partial reactions of protein synthesis except for an approximately sevenfold decrease

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service