Skip to Content
Merck
All Photos(2)

Key Documents

A56450

Sigma-Aldrich

1-Aminohomopiperidine

95%

Synonym(s):

N-Aminohexamethyleneimine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€152.00

€152.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€152.00

About This Item

Empirical Formula (Hill Notation):
C6H14N2
CAS Number:
Molecular Weight:
114.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
95%

€152.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

95%

form

liquid

refractive index

n20/D 1.485 (lit.)

bp

165 °C (lit.)

density

0.984 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

NN1CCCCCC1

InChI

1S/C6H14N2/c7-8-5-3-1-2-4-6-8/h1-7H2

InChI key

UGBKOURNNQREPE-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

132.8 °F - closed cup

Flash Point(C)

56 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Battistina Asproni et al.
Chemical biology & drug design, 91(1), 181-193 (2017-07-05)
Novel 1,4-dihydropyrazolo[3,4-a]pyrrolizine-, 4,5-dihydro-1H-pyrazolo[4,3-g]indolizine- and 1,4,5,6-tetrahydropyrazolo[3,4-c]pyrrolo[1,2-a]azepine-3-carboxamide-based compounds were designed and synthesized for cannabinoid CB

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service