Skip to Content
Merck
All Photos(2)

Key Documents

A55799

Sigma-Aldrich

1-Amino-9-fluorenone

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€293.00

€293.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€293.00

About This Item

Empirical Formula (Hill Notation):
C13H9NO
CAS Number:
Molecular Weight:
195.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€293.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

form

powder

mp

118-120 °C (lit.)

SMILES string

Nc1cccc2-c3ccccc3C(=O)c12

InChI

1S/C13H9NO/c14-11-7-3-6-9-8-4-1-2-5-10(8)13(15)12(9)11/h1-7H,14H2

InChI key

KSEPMOMKAQKOSM-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Thangadurai T et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 199, 465-471 (2017-11-21)
To sense biologically important entities with different size and dimensions, a fluorenone based fluorescent receptor was designed and synthesized. Probe 1 displayed a distinct fluorescence enhancement emission at 565nm for pyrophosphate and 530nm for alanine in polar solvent. The fluorescence

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service