Skip to Content
Merck
All Photos(1)

Key Documents

A54601

Sigma-Aldrich

2-(2-Aminoethyl)isothiourea dihydrobromide

99%

Synonym(s):

2-(2-Aminoethyl)-2-thiopseudourea dihydrobromide, AET, S-(2-Aminoethyl)isothiouronium bromide hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2SC(=NH)NH2 · 2HBr
CAS Number:
Molecular Weight:
281.01
Beilstein:
3911163
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

99%

mp

190-196 °C (lit.)

SMILES string

Br[H].Br[H].NCCSC(N)=N

InChI

1S/C3H9N3S.2BrH/c4-1-2-7-3(5)6;;/h1-2,4H2,(H3,5,6);2*1H

InChI key

XDVMCVGTDUKDHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Has radioprotective activity against x-rays.

Biochem/physiol Actions

Inhibits constitutive and inducible NOS.

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S K Basu et al.
Indian journal of experimental biology, 31(10), 837-842 (1993-10-01)
Radioprotective effectiveness has been evaluated by 30 day survival studies and protection to bone-marrow cells in mice after radiation exposure and this has been further established by 24 hr deoxycytidine excretion in urine of rats following 5 Gy whole body
S K Basu et al.
Indian journal of experimental biology, 30(6), 478-481 (1992-06-01)
Differential radioprotection between normal tissues and carcinoma was observed in C3H/J mice treated with a combination of 5-hydroxy L-tryptophan (5-HTP, 100 mg/kg) and 2-aminoethylisothiuronium bromide hydrobromide (AET, 20 mg/kg). Protection to normal tissues was judged by LD50(30) and by radiation
A Kis et al.
Journal of molecular and cellular cardiology, 31(6), 1229-1241 (1999-06-18)
Right ventricular pacing in lightly anaesthetized dogs (4x5 min periods at a pacing rate of 220 beats/min) protects against the consequences of coronary artery occlusion when this is initiated 24 h after the pacing stimulus. The main purpose of the
Y Gundersen et al.
Shock (Augusta, Ga.), 8(5), 368-372 (1997-11-15)
Endotoxin has profound effects on nitric oxide (NO) production, and considerable controversies exist as to whether these alterations are beneficial or deleterious. Increased mortality has been reported from nonselective inhibition of NO synthase. Results from selective inhibition of the inducible
Ni Ni Tun et al.
Plant & cell physiology, 47(3), 346-354 (2006-01-18)
In this study, we examined the regulation by putrescine, spermidine and spermine of nitric oxide (NO) biosynthesis in Arabidopsis thaliana seedlings. Using a fluorimetric method employing the cell-impermeable NO-binding dye diaminorhodamine-4M (DAR-4M), we observed that the polyamines (PAs) spermidine and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service