96840
H-Lys(Z)-OH
≥99.0% (NT), for peptide synthesis
Synonym(s):
Nε-Z-L-lysine, N6-Carbobenzyloxy-L-lysine
Select a Size
Select a Size
About This Item
Recommended Products
Product Name
H-Lys(Z)-OH, ≥99.0% (NT)
Assay
≥99.0% (NT)
form
powder
optical activity
[α]20/D +15.5±1°, c = 1% in 1 M HCl
reaction suitability
reaction type: solution phase peptide synthesis
mp
259 °C (dec.) (lit.)
application(s)
peptide synthesis
SMILES string
N[C@@H](CCCCNC(=O)OCc1ccccc1)C(O)=O
InChI
1S/C14H20N2O4/c15-12(13(17)18)8-4-5-9-16-14(19)20-10-11-6-2-1-3-7-11/h1-3,6-7,12H,4-5,8-10,15H2,(H,16,19)(H,17,18)/t12-/m0/s1
InChI key
CKGCFBNYQJDIGS-LBPRGKRZSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service