Skip to Content
Merck
All Photos(1)

Key Documents

94070

Sigma-Aldrich

2-Undecanol

≥98.0% (GC)

Synonym(s):

Methyl nonyl carbinol

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€591.00

€591.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€591.00

About This Item

Linear Formula:
CH3(CH2)8CH(OH)CH3
CAS Number:
Molecular Weight:
172.31
Beilstein:
1719795
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€591.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98.0% (GC)

form

liquid

refractive index

n20/D 1.437 (lit.)
n20/D 1.437

bp

130-131 °C/28 mmHg (lit.)
229-231 °C (lit.)

density

0.828 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CCCCCCCCCC(C)O

InChI

1S/C11H24O/c1-3-4-5-6-7-8-9-10-11(2)12/h11-12H,3-10H2,1-2H3

InChI key

XMUJIPOFTAHSOK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

206.6 °F - closed cup

Flash Point(C)

97 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruixia Hong et al.
Journal of separation science, 43(14), 2773-2783 (2020-04-20)
The analysis of plant growth regulators presents a challenge due to their trace quantities and complex matrices. A novel, simple, and effective analytical method for the determination of three trace acidic plant growth regulators in Anoectochilus roxburghii (Wall.) Lindl was
Mohammad Asadi et al.
Journal of separation science, 38(14), 2510-2516 (2015-05-09)
A novel and simple method based on solidified floating organic drop microextraction followed by high-performance liquid chromatography with ultraviolet detection has been developed for simultaneous preconcentration and determination of phenobarbital, lamotrigine, and phenytoin in human plasma and urine samples. Factors

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service