Skip to Content
Merck
All Photos(1)

Documents

939390

Sigma-Aldrich

Cobalt(II) fluoride tetrahydrate

new

99.9% trace metals basis

Synonym(s):

Cobalt difluoride hydrate, Cobaltous fluoride tetrahydrate, Difluorocobalt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CoF2 · 4H2O
CAS Number:
Molecular Weight:
168.99
MDL number:

type

(High purity Salts)

Quality Level

Assay

98-102% (complexometric)
99.9% trace metals basis

form

powder or crystals

impurities

≤1500 ppm (trace metals analysis)

color

faint red to dark red/pink

mp

200 °C

solubility

water: solubleslightly

density

2.19 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤20 ppm

cation traces

Al: <20 ppm
Cu: <20 ppm
Fe: <20 ppm
K: <20 ppm
Mg: <20 ppm
Na: <20 ppm
Pb: <20 ppm

suitability

catalyst

application(s)

(Catalyst)

SMILES string

O.O.O.O.F[Co]F

InChI

1S/Co.2FH.4H2O/h;2*1H;4*1H2/q+2;;;;;;/p-2

InChI key

MNHUTFNEBYCZAF-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Cobalt(II) fluoride tetrahydrate is a reddish crystalline solid with slight solubility in water. It serves as a source of cobalt for oxygen-sensitive applications. Additionally, it is employed as a catalyst biaryl cross-coupling reactions.

Application

Cobalt(II) fluoride tetrahydrate is used as a starting material to synthesize 2-dimensional cobalt-fluoride-oxide as an oxygen evolution electrocatalysts using a rapid thermal annealing (RTA) method. In this process, the cobalt(II) fluoride tetrahydrate is placed in an alumina furnace at 400°C. After annealing for 20 minutes in air, the alumina crucible is removed and allowed to cool naturally to room temperature. The resulting products are then rinsed multiple times in deionized water and ethanol to remove any soluble impurities. Finally, the product is dried thoroughly in a vacuum oven at 60°C to obtain the 2D nanosheets sample. Furthermore, when combined with N-Heterocyclic Carbenes, Cobalt(II) fluoride tetrahydrate has been found to be highly effective as a catalyst for cross-coupling reactions between aryl Grignard reagents and aryl as well as heteroaryl halides. This reaction results in the formation of unsymmetrical biaryls.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid Thermal Annealing toward High-Quality 2D Cobalt Fluoride Oxide as an Advanced Oxygen Evolution Electrocatalyst
Huang, K. t al.
ACS sustainable chemistry & engineering, 8, 6905?6913-6905?6913 (2020)
Takuji Hatakeyama et al.
Journal of the American Chemical Society, 131(33), 11949-11963 (2009-07-31)
Combinations of N-heterocyclic carbenes (NHCs) and fluoride salts of the iron-group metals (Fe, Co, and Ni) have been shown to be excellent catalysts for the cross-coupling reactions of aryl Grignard reagents (Ar(1)MgBr) with aryl and heteroaryl halides (Ar(2)X) to give

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service