Skip to Content
Merck
All Photos(1)

Key Documents

938181

Sigma-Aldrich

Silica

hollow nanoparticles, 1000 nm particle size, pore size 40 Å

Synonym(s):

Hollow silica, Hollow silica nanospheres, Hollow silicon dioxide, Silicon dioxide

Sign Into View Organizational & Contract Pricing

Select a Size

938181-2G
€389.00

€389.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
938181-2G
€389.00

About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
MDL number:
UNSPSC Code:
12352309

€389.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

powder

particle size

1000 nm

pore size

40 Å pore size

bp

2230 °C (lit.)

mp

>1600 °C (lit.)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Our hollow silica nanoparticles are precisely engineered spherical particles of silicon dioxide with a purity of 99.999%. These particles have a uniform particle size of 1000 nm comprised of a porous silica shell, with an average pore size of 40 Å, that surrounds the central hollow cavity. The structure and design of these nanoparticles play a vital role in determining their unique physical characteristics. These properties include low density, a large specific surface area, low dielectric constant, low electrical and thermal conductivity, as well as abnormal light scattering behavior.

Application

Hollow silica nanoparticles find application in many areas including drug delivery, catalysis, thermal insulation, and functional coatings. In drug delivery, the hollow interior of the nanoparticles can be used to encapsulate and deliver drugs to specific targets.[1] The average 2-nm sized pores can selectively allow certain molecules to pass through while blocking or slowing others. In catalysis, the nanoparticles can act as catalyst supports due to their high surface area and porosity. Hollow silica nanoparticles impregnated with the catalyst may be particularly useful in environments with complex matrices where the porosity of the particle facilitates the isolation of reactants from matrix elements. In electronic applications, hollow silica’s low thermal conductivity can be utilized to improve thermal insulation properties and their monodisperse particle size facilitates their self-assembly into arrays with unique optical and electrical properties.[2]

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hollow Silica Particles: Recent Progress and Future Perspectives
Sharma J, et al.
Nanomaterials (Basel, Switzerland), 10, 1599-1599 (2020)
A Comprehensive Study of Drug Loading in Hollow Mesoporous Silica Nanoparticles: Impacting Factors and Loading Efficiency
Guo L, et al.
Nanomaterials (Basel, Switzerland), 11, 1293-1293 (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service