Skip to Content
Merck
All Photos(1)

Documents

93479

Sigma-Aldrich

Chloromethanesulfonyl chloride

technical, ≥90% (AT)

Synonym(s):

Chloromesyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2SO2Cl
CAS Number:
Molecular Weight:
149.00
Beilstein:
1811761
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥90% (AT)

form

liquid

density

1.64 g/mL at 20 °C (lit.)

storage temp.

−20°C

SMILES string

ClCS(Cl)(=O)=O

InChI

1S/CH2Cl2O2S/c2-1-6(3,4)5/h1H2

InChI key

KQDDQXNVESLJNO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service