Skip to Content
Merck
All Photos(1)

Key Documents

934135

Sigma-Aldrich

TissueFab® Sodium Persulfate Solution

low endotoxin

Synonym(s):

Sodium persulfate, SPS, SPS Crosslinker, Sodium peroxodisulfate

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€44.80

€44.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
€44.80

About This Item

Linear Formula:
Na2S2O8
CAS Number:
Molecular Weight:
238.10
MDL number:
UNSPSC Code:
12352300
NACRES:
NA.21

€44.80


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[O-]S(=O)(=O)OOS([O-])(=O)=O

InChI

1S/2Na.H2O8S2/c;;1-9(2,3)7-8-10(4,5)6/h;;(H,1,2,3)(H,4,5,6)/q2*+1;/p-2

InChI key

CHQMHPLRPQMAMX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Rutheneum (Ru) and sodium persulfate (SPS) have the ability to effectively generate free radicals in the 400-450nm visible light range, allowing for cell-friendly photo-crosslinking of a variety of hydrogels[1]. Among the materials that can be crosslinked via this method are the natturally derived and biocompatible biomaterials silk fibroin and gelatin[2] . These materials can be used to enhance the mechanical and rheological properties to aid the 3D bioprinting workflow.

Application

Sodium persulfate is provided at an optimized concentration for 3D bioprinting applications. It can be used to bioprint via extrusion based 3D bioprinting cell-laden hydrogels in desired shapes and without any supporting materials. Crosslinking of the 3D printed structures is done in one step using visible light. The crosslinked cell-laden bioinks can be further culutred and use for tissue engineering and regenerative medicina applications.
Low Endotoxin, low bioburden: Endotoxins have been demonstrated negatively impact cellular growth, morphology, differentiation, inflammation and protein expression. Bioburden is defined as the number of contaminated organisms found in a given amount of material. We test each lot for endotoxins as well as total bioburden (aerobic and fungal) to minimize unwanted interactions. For more information: https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/microbiological-testing/pyrogen-testing/what-is-endotoxin

Legal Information

Tissuefab is a registered trademark of Sigma-Aldrich Co. LLC
TISSUEFAB is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame over circleHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Ox. Liq. 3 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Khoon S Lim et al.
Macromolecular bioscience, 19(6), e1900098-e1900098 (2019-04-27)
In this study, the cyto-compatibility and cellular functionality of cell-laden gelatin-methacryloyl (Gel-MA) hydrogels fabricated using a set of photo-initiators which absorb in 400-450 nm of the visible light range are investigated. Gel-MA hydrogels cross-linked using ruthenium (Ru) and sodium persulfate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service