Skip to Content
Merck
All Photos(1)

Documents

934127

Sigma-Aldrich

TissueFab® bioink SilkGel -Vis/405nm SPS crosslinking kit, low endotoxin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352201
NACRES:
NA.28

description

low endotoxin

Quality Level

storage temp.

2-8°C

General description

The low endotoxin TissueFab® bioink SilkGel -Vis/405nm SPS crosslinking kit can be used to bioprint cell-laden hydrogels in the desired shape without any supporting material. This bioink can be crosslinked in one step using exposure to 405 nm light for further culture and maturation of cells for tissue engineering and regenerative medicine applications.

Application

Currently, there is a need for high-quality, commercially available ready-to-use bioink formulations to enable the reproducible fabrication of synthetic tissues and organs by 3D Bioprinting
  • drug discovery
  • in-vitro disease models
  • regenerative medicine-cell
  • cultured meat

Features and Benefits

  • Bioprinting models replicate biology for drug discovery and in vivo applications
  • Sterile, low endotoxin
  • Batch control offers reproducible models for preclinical toxicology testing and drug screening
  • Extended shelf-life & stability

Legal Information

TISSUEFAB is a registered trademark of Merck KGaA, Darmstadt, Germany

Kit Components Only

Product No.
Description

  • TissueFab® bioink SilkGel -Vis/405nm, low endotoxin

  • TissueFab® Sodium Persulfate Solution, low endotoxin

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Ox. Liq. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Khoon S Lim et al.
Macromolecular bioscience, 19(6), e1900098-e1900098 (2019-04-27)
In this study, the cyto-compatibility and cellular functionality of cell-laden gelatin-methacryloyl (Gel-MA) hydrogels fabricated using a set of photo-initiators which absorb in 400-450 nm of the visible light range are investigated. Gel-MA hydrogels cross-linked using ruthenium (Ru) and sodium persulfate
Soon Hee Kim et al.
Nature communications, 9(1), 1620-1620 (2018-04-26)
Although three-dimensional (3D) bioprinting technology has gained much attention in the field of tissue engineering, there are still several significant engineering challenges to overcome, including lack of bioink with biocompatibility and printability. Here, we show a bioink created from silk

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service