Skip to Content
Merck
All Photos(2)

Key Documents

933988

Sigma-Aldrich

1,1,2,2-Tetrafluoroethyl 2,2,2-trifluoroethyl ether

greener alternative

≥99.5%, anhydrous, acid <=100 ppm, battery grade

Synonym(s):

1,1,2,2-tetrafluoro-1-(2,2,2-trifluoroethoxy)ethane, HFE-347, TFTFE

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€293.00

€293.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€293.00

About This Item

Empirical Formula (Hill Notation):
C4H3F7O
CAS Number:
Molecular Weight:
200.05
Enzyme Commission number:
609-858-6
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

€293.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

battery grade

Quality Level

Assay

≥99.5%

form

liquid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤100 ppm acid (HF)
≤250 ppm H2O

non-volatile residue (NVR)

≤10 ppm

bp

56 °C

mp

-91 °C (lit.)

density

1.49 g/mL

application(s)

battery manufacturing

greener alternative category

SMILES string

FC(F)(F)COC(F)(F)C(F)F

InChI

1S/C4H3F7O/c5-2(6)4(10,11)12-1-3(7,8)9/h2H,1H2

InChI key

CWIFAKBLLXGZIC-UHFFFAOYSA-N

General description

1,1,2,2-Tetrafluoroethyl 2,2,2-trifluoroethyl ether (TFTFE) is a fluorinated ether that finds extensive use as an electrolyte solvent and diluent in various battery technologies. TFTFE has a low viscosity, low freezing point (-94 °C lit.), low dielectric constant (~6.7), and high electrochemical stability, making it an ideal candidate for use in lithium-ion batteries, lithium-sulfur batteries, and other battery systems. TFTFE is miscible with many polar organic solvents, including carbonates typically used in battery electrolytes. With a minimum purity level of 99% and free from acid impurities, our TFTFE is a reliable and safe solution for critical battery applications.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

Battery-grade 1,1,2,2-tetrafluoroethyl 2,2,2-trifluoroethyl ether (TFTFE) is a versatile co-solvent and additive for various battery systems. In lithium-metal batteries, TFTFE helps to suppress dendrites without raising the interfacial impedance. It also supports the stable cycling of NMC and lithium metal phosphate cathodes by forming a highly fluorinated interphase, which inhibits oxidation and transition metal dissolution.[1] Because of its stability and low viscosity, TFTFE is commonly added in localized high-concentration electrolytes (LHCE) as a diluent and flame-retardant.[2] In lithium-sulfur batteries, TFTFE plays a key role as both a polysulfide-restraining solvent and a film-forming agent, addressing the polysulfide shuttle (PSS) effect and improving battery performance.[3][4] Additionally, TFTFE plays a critical role in cell systems with solvate ionic liquids (SIL) as an ionic conduction-enhancing ingredient, particularly for high-rate cycle environments.[5] Our high-purity, anhydrous TFTFE is an ideal battery-grade additive for advanced battery technology.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solvate ionic liquid electrolyte with 1,1,2,2-tetrafluoroethyl 2,2,2-trifluoroethyl ether as a support solvent for advanced lithium?sulfur batteries
Lu, Hai, et al.
Royal Society of Chemistry Advances, 6, 18186-18190 (2016)
Application of Partially Fluorinated Ether for Improving Performance of Lithium/Sulfur Batteries
Lu, Hai, et al.
Journal of the Electrochemical Society, 162, A1460-A1460 (2015)
Synthesis and electrochemical properties of partially fluorinated ether solvents for lithiumsingle bondsulfur battery electrolytes
Yue Zheng
Journal of Power Sources, 401, 271-277 (2018)
Jun-Fan Ding et al.
Angewandte Chemie (International ed. in English), 60(20), 11442-11447 (2021-03-04)
Lithium (Li) metal anodes hold great promise for next-generation high-energy-density batteries, while the insufficient fundamental understanding of the complex solid electrolyte interphase (SEI) is the major obstacle for the full demonstration of their potential in working batteries. The characteristics of
Xiulin Fan et al.
Nature nanotechnology, 13(8), 715-722 (2018-07-18)
Rechargeable Li-metal batteries using high-voltage cathodes can deliver the highest possible energy densities among all electrochemistries. However, the notorious reactivity of metallic lithium as well as the catalytic nature of high-voltage cathode materials largely prevents their practical application. Here, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service