Skip to Content
Merck
All Photos(3)

Key Documents

933767

Sigma-Aldrich

Silicon

greener alternative

nanoparticles, 40 nm avg. part. size, ≥99% trace metals basis

Synonym(s):

SiNPs, Silicon nanopowder

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€391.00

€391.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€391.00

About This Item

Linear Formula:
Si
CAS Number:
Molecular Weight:
28.09
MDL number:
UNSPSC Code:
12352309
NACRES:
NA.21

€391.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

battery grade

Quality Level

description

Application: Battery Manufacturing

Assay

≥99% trace metals basis

form

nanoparticles
powder or crystals

composition

(Si)

storage condition

(Avoid contact with skin and eyes. Keep container tightly closed in a dry and well-ventilated place.)

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

avg. part. size

40 nm

bp

2355 °C (lit.)

mp

1410 °C (lit.)

density

2.33 g/mL at 25 °C (lit.)

bulk density

0.07‑0.08 g/cm3

application(s)

battery manufacturing

greener alternative category

SMILES string

[Si]

InChI

1S/Si

InChI key

XUIMIQQOPSSXEZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Our 40 nm silicon nanoparticles are a high-quality, ultrafine powder with a narrow particle size of 40 nm and a high surface area (~100 m2/g). It is produced by the laser gas phase synthetic method from high-purity silane to achieve the high purity (>9=9%) and reliable particle size.
We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives. This product has been enhanced for energy efficency. Click here for more information.

Application

Silicon nanopowder is a promising material for use in advanced battery technologies. With its high theoretical capacity (4212 mAh/g), it has the potential to significantly improve the performance of lithium-ion batteries. The controlled particle size and high surface area of our silicon nanopowder make it an ideal candidate for use as an anode material in these batteries, allowing for improved charge and discharge rates, longer cycle life, and higher energy densities. Additionally, silicon nanopowder is being researched for use in other types of batteries, including lithium-air and lithium-sulfur batteries. Its unique properties make it a valuable material for developing next-generation battery technologies that offer higher performance, longer lifetimes, and lower costs.

Features and Benefits

Our silicon nanoparticles are designed for battery applications to extend cycle life and charge and discharge rates. We offer:
  • Low Particle Size
  • High Surface Area
  • Consistent Quality

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rational design of silicon-based composites for high-energy storage devices
Lee, Jung Kyoo, et al.
Journal of Materials Chemistry, 4, 5366- 5384 (2016)
Vanitha Selvarajan et al.
Frontiers in chemistry, 8, 602-602 (2020-08-08)
The rapid emergence of drug resistance continues to outpace the development of new antibiotics in the treatment of infectious diseases. Conventional therapy is currently limited by drug access issues such as low intracellular drug accumulations, drug efflux by efflux pumps

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service