933244
FIB holder
Double 1/2" FIB Sample and Grid Holder, Pin
Synonym(s):
FIB mount
Select a Size
Select a Size
About This Item
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Pin: Standard 3.2 mm (⅛").
Material: Vacuum grade aluminum with brass screws.
Includes Philips screwdriver #0.
Application
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service