Skip to Content
Merck
All Photos(1)

Key Documents

929360

Sigma-Aldrich

Opto-pomalidomide

≥95%

Synonym(s):

4,5-Dimethoxy-2-nitrobenzyl 3-(4-amino-1,3-dioxoisoindolin-2-yl)-2,6-dioxopiperidine-1-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€287.00

€287.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€287.00

About This Item

Empirical Formula (Hill Notation):
C23H20N4O10
Molecular Weight:
512.43
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.21

€287.00


Please contact Customer Service for Availability

Request a Bulk Order

ligand

pomalidomide

Quality Level

Assay

≥95%

form

powder

storage temp.

2-8°C

SMILES string

NC1=C2C(C(N(C2=O)C3C(N(C(CC3)=O)C(OCC4=C(C=C(C(OC)=C4)OC)[N+]([O-])=O)=O)=O)=O)=CC=C1

InChI

1S/C23H20N4O10/c1-35-16-8-11(15(27(33)34)9-17(16)36-2)10-37-23(32)26-18(28)7-6-14(21(26)30)25-20(29)12-4-3-5-13(24)19(12)22(25)31/h3-5,8-9,14H,6-7,10,24H2,1-2H3

InChI key

VIMWIXPVAGZBOD-UHFFFAOYSA-N

Application

Protein degrader building block Opto-pomalidomide enables the synthesis of molecules for light-induced targeted protein degradation and PROTAC® (proteolysis-targeting chimeras) research. This conjugate contains a Cereblon (CRBN) recruiting ligand, a rigid linker, and a nitroveratryloxycarbonyl group that can undergo photolysis to form a pendant amine for reactivity with a carboxylic acid on the target ligand. Because even slight alterations in ligands and crosslinkers can affect ternary complex formation between the target, E3 ligase, and degrader, many analogs are prepared to screen for optimal target degradation. When used with other protein degrader building blocks with a terminal amine, parallel synthesis can be used to more quickly generate degrader libraries that feature variation in crosslinker length, composition, and E3 ligase ligand.


Technology Spotlight: Degrader Building Blocks for Targeted Protein Degradation


Protein Degrader Building Blocks

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Liu et al.
Science advances, 6(8), eaay5154-eaay5154 (2020-03-05)
By hijacking endogenous E3 ligase to degrade protein targets via the ubiquitin-proteasome system, PROTACs (PRoteolysis TArgeting Chimeras) provide a new strategy to inhibit protein targets that were regarded as undruggable before. However, the catalytic nature of PROTAC potentially leads to
Daniel P Bondeson et al.
Annual review of pharmacology and toxicology, 57, 107-123 (2016-10-13)
Protein homeostasis networks are highly regulated systems responsible for maintaining the health and productivity of cells. Whereas therapeutics have been developed to disrupt protein homeostasis, more recently identified techniques have been used to repurpose homeostatic networks to effect degradation of
Kedra Cyrus et al.
Molecular bioSystems, 7(2), 359-364 (2010-10-06)
Conventional genetic approaches have provided a powerful tool in the study of proteins. However, these techniques often preclude selective manipulation of temporal and spatial protein functions, which is crucial for the investigation of dynamic cellular processes. To overcome these limitations
Momar Toure et al.
Angewandte Chemie (International ed. in English), 55(6), 1966-1973 (2016-01-13)
The current inhibitor-based approach to therapeutics has inherent limitations owing to its occupancy-based model: 1) there is a need to maintain high systemic exposure to ensure sufficient in vivo inhibition, 2) high in vivo concentrations bring potential for off-target side effects, and 3) there is
Philipp M Cromm et al.
Cell chemical biology, 24(9), 1181-1190 (2017-06-27)
Traditional pharmaceutical drug discovery is almost exclusively focused on directly controlling protein activity to cure diseases. Modulators of protein activity, especially inhibitors, are developed and applied at high concentration to achieve maximal effects. Thereby, reduced bioavailability and off-target effects can

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service