Skip to Content
Merck
All Photos(1)

Key Documents

929298

Sigma-Aldrich

ItOct HCl

≥95.0%

Synonym(s):

1,3-Bis(2,4,4-trimethylpentan-2-yl)-1H-imidazol-3-ium chloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€88.30
500 MG
€243.00

€88.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€88.30
500 MG
€243.00

About This Item

Empirical Formula (Hill Notation):
C19H37ClN2
Molecular Weight:
328.96
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.21

€88.30


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥95.0%

form

powder

parameter

moisture stable

SMILES string

CC(C)(CC(C)(C)C)N1C=[N+](C(C)(CC(C)(C)C)C)C=C1.[Cl-]

InChI

1S/C19H37N2.ClH/c1-16(2,3)13-18(7,8)20-11-12-21(15-20)19(9,10)14-17(4,5)6;/h11-12,15H,13-14H2,1-10H3;1H/q+1;/p-1

InChI key

USCUVFCUVMJDDA-UHFFFAOYSA-M

Application

ItOct HCl is an N-alkyl NHC ligand with demonstrated uses in the following reactions. Au-catalyzed hydration, Cu-catalyzed hydroboration, Ag-catalyzed hydroboration, Cu-catalyzed C–O coupling and Pd-catalyzed C–C and C–N couplings. This ligand features significantly higher steric volume compared with ItBu (659983) while maintaining the σ-donation and σ-acceptance inherent to N-alkyl NHC ligands.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service