928437
TissueFab® bioink Facile Curable Gel
Cationic Crosslinking
Synonym(s):
3D Bioprinting, Bioink, Facile Curable
Select a Size
Select a Size
About This Item
Recommended Products
Quality Level
sterility
0.2 μm filtered
form
viscous liquid (or gel)
composition
Cationic/ionic
Gelatin
impurities
<5 CFU/g Bioburden(Total aerobic)
<5 CFU/g Bioburden(fungal)
color
pale yellow to colorless
viscosity
2-20 cP
compatibility
3D bioprinting, extrusion bioprinting
General description
The bioink has a Gelatin base which as a natural protein offers excellent cytocompatibility and cell-binding motifs in order to mimic the native extracellular microenvironment.
Low Endotoxin, low bioburden: Endotoxins have been demonstrated negatively impact cellular growth, morphology, differentiation, inflammation and protein expression. Bioburden is defined as the number of contaminated organisms found in a given amount of material. We test each lot for endotoxins as well as total bioburden (aerobic and fungal) to minimize unwanted interactions. For more information: https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/microbiological-testing/pyrogen-testing/what-is-endotoxin
Packaging
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service