Skip to Content
Merck
All Photos(2)

Key Documents

92759

Sigma-Aldrich

tert-Butyl trimethylsilylacetate

≥95.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H20O2Si
CAS Number:
Molecular Weight:
188.34
Beilstein:
1930256
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (GC)

form

liquid

refractive index

n20/D 1.417
n20/D 1.418 (lit.)

bp

67 °C/13 mmHg (lit.)

density

0.86 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CC(C)(C)OC(=O)C[Si](C)(C)C

InChI

1S/C9H20O2Si/c1-9(2,3)11-8(10)7-12(4,5)6/h7H2,1-6H3

InChI key

HOHBQWITMXOSOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent for the synthesis of α,β-unsaturated esters[1]; Formation of β-keto esters[2][3]

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

107.6 °F - closed cup

Flash Point(C)

42 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S.L. Hartzell et al.
Tetrahedron Letters, 2737-2737 (1976)
S.L. Hartzell et al.
Tetrahedron Letters, 1403-1403 (1974)
Tetrahedron Letters, 2757-2757 (1976)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service