Skip to Content
Merck
All Photos(1)

Key Documents

926698

Sigma-Aldrich

MCM-41 100nm nanoparticle

pore size 30 Å, 1% in water (dispersion)

Synonym(s):

Mesoporous silica, Silica nanoparticles

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
€257.00

€257.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
2 ML
€257.00

About This Item

Linear Formula:
SiO2
CAS Number:
MDL number:
UNSPSC Code:
12352309
NACRES:
NA.22

€257.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

concentration

1% in water (dispersion)

pore size

30 Å pore size

SMILES string

[Si](=O)=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silica, mesostructured (MCM-41) is a part of low-density organized mesoporous silicate system which has a 2D hexagonal phase. MCM-41 nanoparticles have a wide range of unique features such as durability, high specific surface area, air tolerance, large and uniform pore size, and high thermal and mechanical stability.

Application

Their applications are consequences of their porous nature and robust chemical composition, which permit their use in many applications:
  • water treatment
  • gas processing
  • supported catalysis
  • drug delivery

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mesoporous Silica Nanoparticles for Bio-Applications.
Niculescu V, et al.
Frontiers in materials, 7 (2020)
Functionalized Ordered Mesoporous Silicas (MCM-41): Synthesis and Applications in Catalysis.
Martinez-Edo G, et al.
Catalysts, 8(12), 617-617 (2018)
Mesoporous MCM-41 material for hydrogen storage: A short review.
Tekkaya E, et al.
International Journal of Hydrogen Energy, 41(23), 9789-9795 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service