923818
N-(But-3-yn-1-yl)-2-(((2-oxo-2H-chromen-7-yl)oxy)methyl)acrylamide
Synonym(s):
Coumarin alkyne CoLDR probe
About This Item
Recommended Products
reaction suitability
reagent type: chemical modification reagent
reaction type: click chemistry
Quality Level
functional group
(Alkyne)
storage temp.
2-8°C
SMILES string
O=C1OC2=CC(OCC(C(NCCC#C)=O)=C)=CC=C2C=C1
InChI key
MTTOLTREJYBDSH-UHFFFAOYSA-N
Related Categories
Application
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service