Skip to Content
Merck
All Photos(1)

Key Documents

920088

Sigma-Aldrich

Graphene, monolayer film

large grain, CVD graphene on copper foil, A5 size

Synonym(s):

Roll-to-Roll graphene

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€919.00

€919.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EACH
€919.00

About This Item

Linear Formula:
C
CAS Number:
MDL number:
UNSPSC Code:
12141908
NACRES:
NA.23

€919.00


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

R2R Monolayer large grain CVD graphene on copper foil, A5 size, avg. no. of layers, 1

Quality Level

description

Growth method: roll-to-roll CVD
Raman intensity 2D/G: ≥1.5
Number of layer: Monolayer

feature

avg. no. of layers 1

sheet resistance

280 Ω/sq ±10%

size

110 μm × 110 μm ± 10%, grain size

thickness

35 μm , Cu Foil

surface coverage

surface coverage >98%

transmittance

>97%

semiconductor properties

(mobility>3000 cm2/V·s) (Hall effect measurements)

SMILES string

NC.NC.NC.NC.NC.NC.NC.NC.NC

InChI

1S/9CH5N/c9*1-2/h9*2H2,1H3

InChI key

DXIFQVRACSPGSU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Roll-to-roll, high-quality, monolayer CVD graphene with large grain size (∼110μm2) on copper foil, A5 size.

Application

Our Roll-to-Roll CVD graphene products are true monolayer high quality graphene, fabricated inside a Cleanroom, heavily monitored and QC to assure high reproducibility.
The roll-to-roll process allows continuous, large scale graphene production.[1]

This large grain size product with low sheet resistance would enable unmatched reproducibility and allow high performance for CVD graphene based FET[2], CVD graphene based sensors, and heterostructure based micro/nano electronics[3].

Application examples:
  • Ultrafast Transistor
  • Optical devices
  • Bio/Gas Sensor
  • Transparent Electrode
  • Flexible Display
  • Smart Coating
  • Thermal management

Caution

Be cautious not to drop
Keep away from contamination, heat, dust and flame etc.

Storage and Stability

Avoid direct sun light, avoid high temperature, avoid high humidity, and avoid dust.

Legal Information

Product of LG Electronics, R&D use only

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Review on 1D vdWHs: Discusses materials, synthesis, optoelectronic applications, challenges, and future perspectives for 1D vdWH-based devices.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service