Skip to Content
Merck
All Photos(3)

Documents

916420

Sigma-Aldrich

BIAN-IPr# HCl

Synonym(s):

7,9-Bis-(2,4,6-Tribenzhydrylphenyl)-7H-acenaphtho[1,2-d]imidazol-9-ium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C103H77ClN2
Molecular Weight:
1378.18

form

powder

Application

BIAN-IPr# HCl is sterically-bulky, easily-accessible NHC ligand that can be used in a broad range of catalytic reactions, including cross-coupling reactions, hydrofunctionalizations and element addition reactions using different metals, including but not limited to Pd, Ni, Au, Ag, Cu, Pt, Ru.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service