Skip to Content
Merck
All Photos(2)

Key Documents

916382

Sigma-Aldrich

NanoFabTx reagent

PCL Nano

Synonym(s):

NanoFabTx

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€460.00

€460.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€460.00

About This Item

Empirical Formula (Hill Notation):
C6H10O2·C2H4O)x·CH4O
CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

€460.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

application(s)

advanced drug delivery

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

NanoFabTx reagent, PCL-Nano is a component of our NanoFabTx nanoparticle formulation screening kit (917737) which includes additional reagents, stabilizer and protocols for nanoprecipitation and microfluidics to synthesize 100-200 nm nanoparticles. NanoFabTx reagent, PCL-Nano is a polycaprolactone (PCL) polymer, which is an FDA approved biodegradable polymer for biomedical and pharmaceutical applications, widely used as drug delivery systems.

Application

NanoFabTx reagent, PCL-Nano can be purchased separately as a refill for our formulation screening kit (917737).

Legal Information

NANOFABTX is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service