Skip to Content
Merck
All Photos(2)

Key Documents

915157

Sigma-Aldrich

iPr-PDI

≥95%

Synonym(s):

N,N-Bis(2,6-diisopropylphenyl)perylene-3,4,9,10-bis(dicarboximide)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€171.00
500 MG
€602.00

€171.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€171.00
500 MG
€602.00

About This Item

Empirical Formula (Hill Notation):
C48H42N2O4
CAS Number:
Molecular Weight:
710.86
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

€171.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

powder

mp

>300 °C

storage temp.

−20°C

SMILES string

[n]2([c](c3c4c5c(c6c7c8c([c]([n]([c](c8cc6)=O)c9c(cccc9C(C)C)C(C)C)=O)ccc7c5cc3)ccc4[c]2=O)=O)c1c(cccc1C(C)C)C(C)C

InChI

1S/C48H42N2O4/c1-23(2)27-11-9-12-28(24(3)4)43(27)49-45(51)35-19-15-31-33-17-21-37-42-38(22-18-34(40(33)42)32-16-20-36(46(49)52)41(35)39(31)32)48(54)50(47(37)53)44-29(25(5)6)13-10-14-30(44)26(7)8/h9-26H,1-8H3

InChI key

NAZODJSYHDYJGP-UHFFFAOYSA-N

Application

iPr-PDI is a perylene based organic photocatalyst with a variety of demonstrated uses, including the reduction of aryl halides and the iodoperfluoroalkylation of alkenes.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cristian Rosso et al.
Organic letters, 21(13), 5341-5345 (2019-06-30)
Two efficient continuous flow iodoperfluoroalkylation methods are described: using 0.05 mol % perylene diimide (PDI) photocatalyst under 450 nm irradiation or substoichiometric triethylamine under 405 nm irradiation. These methods enable dramatically elevated productivity versus batch processes. The triethylamine-mediated method is
Indrajit Ghosh et al.
Science (New York, N.Y.), 346(6210), 725-728 (2014-11-08)
Biological photosynthesis uses the energy of several visible light photons for the challenging oxidation of water, whereas chemical photocatalysis typically involves only single-photon excitation. Perylene bisimide is reduced by visible light photoinduced electron transfer (PET) to its stable and colored

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service