Skip to Content
Merck
All Photos(1)

Key Documents

914908

Sigma-Aldrich

Lithium modified silica nanoparticles

pore size non-porous, 200 nm particle size, 20 % (w/w) in ethanol

Synonym(s):

Battery electrolytes, Nanosalt, Silica functionalized li-ion particles

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€280.00

€280.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€280.00

About This Item

Linear Formula:
SiO2O(CH2)3SO3-Li+
CAS Number:
MDL number:
UNSPSC Code:
12352300
NACRES:
NA.23

€280.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

battery grade

Quality Level

Assay

≥95%

form

liquid

concentration

20 % (w/w) in ethanol

particle size

200 nm

pore size

non-porous

application(s)

battery manufacturing

SMILES string

[Si](=O)=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Monodisperse nonporous silica nanoparticles with lithium ion functionalization have been used in electrolytes and nanosalts [[1][2][3]]. We offer highly monodisperse nonporous silica particles with lithium tertbutoxide, with sizes varying from 200 nm to 600 nm.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

56.3 °F

Flash Point(C)

13.5 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gel electrolytes based on lithium modified silica nano-particles.
Sun J, et al.
Electrochimica Acta, 52, 708-7090 (2007)
Supercapacitive properties of activated carbon electrode in electrolyte solution with a lithium-modified silica nanosalt.
Jung H W, et al.
Current Applied Physics, 15, 567-570 (2015)
Lithium modified silica as electrolyte additive for lithium secondary batteries.
Latifatu M, et al.
Solid State Ionics, 319, 7-12 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service