Skip to Content
Merck
All Photos(2)

Key Documents

914789

Sigma-Aldrich

iPr-NDI

≥95%

Synonym(s):

Isopropyl naphthyridine-diimine

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€306.00

€306.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€306.00

About This Item

Empirical Formula (Hill Notation):
C36H44N4
CAS Number:
Molecular Weight:
532.76
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

€306.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

powder

mp

266-269 °C

SMILES string

N(c4c(cccc4C(C)C)C(C)C)C(=C)c1nc2nc(ccc2cc1)C(=C)Nc3c(cccc3C(C)C)C(C)C

InChI

1S/C36H44N4/c1-21(2)28-13-11-14-29(22(3)4)34(28)37-25(9)32-19-17-27-18-20-33(40-36(27)39-32)26(10)38-35-30(23(5)6)15-12-16-31(35)24(7)8/h11-24,37-38H,9-10H2,1-8H3

InChI key

VLOKSQBHSIFTBM-UHFFFAOYSA-N

Application

iPr-NDI is a ligand that when mixed with NiBr2, generates an active dinuclear Ni catalyst. The catalyst has been shown to promote a variety of transformations including the cyclopropanation of alkenes and the [4+1] cycloaddition of vinylidenes and dienes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ian G Powers et al.
Journal of the American Chemical Society, 140(11), 4110-4118 (2018-03-01)
Azoarenes are valuable chromophores that have been extensively incorporated as photoswitchable elements in molecular machines and biologically active compounds. Here, we report a catalytic nitrene dimerization reaction that provides access to structurally and electronically diverse azoarenes. The reaction utilizes aryl
You-Yun Zhou et al.
Angewandte Chemie (International ed. in English), 55(9), 3171-3175 (2016-01-30)
Dinuclear Ni complexes supported by naphthyridine-diimine (NDI) ligands catalyze the reductive cyclopropanation of alkenes with CH2 Cl2 as the methylene source. The use of mild terminal reductants (Zn or Et2 Zn) confers significant functional-group tolerance, and the catalyst accommodates structurally
Douglas R Hartline et al.
Journal of the American Chemical Society, 139(39), 13672-13675 (2017-09-19)
Single bonds between carbon atoms are inherently challenging to activate using transition metals; however, ring-strain release can provide the necessary thermodynamic driving force to make such processes favorable. In this report, we describe a strain-induced C-C oxidative addition of norbornadiene.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service