Skip to Content
Merck
All Photos(1)

Key Documents

914487

Sigma-Aldrich

2-(Diphenylphosphanyl)benzamide

≥95%

Synonym(s):

2-(Diphenylphosphino)-benzamide, DPBM, Phosphine for Staundinger reduction

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€101.00
250 MG
€312.00

€101.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€101.00
250 MG
€312.00

About This Item

Empirical Formula (Hill Notation):
C19H16NOP
CAS Number:
Molecular Weight:
305.31
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22
form:
powder
Assay:
≥95%

€101.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

powder

storage temp.

−20°C

SMILES string

P(c3c(cccc3)C(=O)N)(c2ccccc2)c1ccccc1

InChI

1S/C19H16NOP/c20-19(21)17-13-7-8-14-18(17)22(15-9-3-1-4-10-15)16-11-5-2-6-12-16/h1-14H,(H2,20,21)

General description

It is recommended to make fresh stock stolutions prior to each use.

Application

2-(Diphenylphosphanyl)benzamide (DPBM) is a phosphine useful in the Staudinger reduction in chemical biology and medicinal chemistry. Recently, DPBM was demonstrated to faciliate the reactivation of CRISPR/Cas9 functions by removing azidomethylnicotinyl (AMN) groups used to masked gRNA and inhibit CRISPR systems. AMN groups were originally added with the use of RNA probe NAI-N3 (cat# 913812).[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shao-Ru Wang et al.
Nature communications, 11(1), 91-91 (2020-01-05)
Prokaryotes use repetitive genomic elements termed CRISPR (clustered regularly interspaced short palindromic repeats) to destroy invading genetic molecules. Although CRISPR systems have been widely used in DNA and RNA technology, certain adverse effects do occur. For example, constitutively active CRISPR

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service